ENA390552867 Enamine

7-bromo-5-fluoro-1,2,3,4-tetrahydroisoquinolin-1-one

  • MDL number MFCD20287127
  • Cas Number : 1341674-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127999
Availability details for
Quantity
  • Availability
  • Add to Cart