ENA390552873 Enamine

methyl 5-chloropentanecarboximidate hydrochloride

  • MDL number MFCD20624416
  • Cas Number : 77570-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128021
Availability details for
Quantity
  • Availability
  • Add to Cart