ENA390552876 Enamine

4-(phenylcarbamoyl)phenyl acetate

  • MDL number MFCD01052611
  • Cas Number : 33577-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128026
Availability details for
Quantity
  • Availability
  • Add to Cart