ENA390552879 Enamine

ethyl 1-methyl-5-phenyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD09028449
  • Cas Number : 116344-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128031
Availability details for
Quantity
  • Availability
  • Add to Cart