ENA390552884 Enamine

6,6-dimethyl-4,5,6,7-tetrahydro-1H-1,3-benzodiazol-4-one

  • MDL number MFCD13811554
  • Cas Number : 21815-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128106

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128106
Availability details for
Quantity
  • Availability
  • Add to Cart