ENA390552887 Enamine

2-(1-chloroethyl)-1,3-benzoxazole

  • MDL number MFCD10691335
  • Cas Number : 1092286-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126602
Availability details for
Quantity
  • Availability
  • Add to Cart