ENA390552894 Enamine

2-(2-iodoethoxy)-2-methylpropane

  • MDL number MFCD24369119
  • Cas Number : 156937-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126619
Availability details for
Quantity
  • Availability
  • Add to Cart