ENA390552895 Enamine

4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]

  • MDL number MFCD19703995
  • Cas Number : 1283095-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119747
Availability details for
Quantity
  • Availability
  • Add to Cart