ENA390552896 Enamine

3-[5-(methanesulfonylmethyl)-1,3,4-oxadiazol-2-yl]benzoic acid

  • MDL number MFCD19564162
  • Cas Number : 1409023-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126633
Availability details for
Quantity
  • Availability
  • Add to Cart