ENA390552897 Enamine

3,4-dihydrospiro[2-benzopyran-1,4'-piperidine]

  • MDL number MFCD16876099
  • Cas Number : 180160-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128483
Availability details for
Quantity
  • Availability
  • Add to Cart