ENA390552899 Enamine

6-tert-butyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2-carbaldehyde

  • MDL number MFCD17251091
  • Cas Number : 1341677-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126637
Availability details for
Quantity
  • Availability
  • Add to Cart