ENA390552903 Enamine

tert-butyl decahydro-1H-1,4-benzodiazepine-4-carboxylate

  • MDL number MFCD22033469
  • Cas Number : 1369133-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126644

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126644
Availability details for
Quantity
  • Availability
  • Add to Cart