ENA390552904 Enamine

(2S)-2-amino-N,N-dimethylpropanamide hydrochloride

  • MDL number MFCD20487760
  • Cas Number : 125218-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126648

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-126648
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart