ENA390552912 Enamine

1-(2,4-difluorophenyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD11037028
  • Cas Number : 1260672-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126672
Availability details for
Quantity
  • Availability
  • Add to Cart