ENA390552925 Enamine

4-chloro-7-methoxy-3-nitroquinoline

  • MDL number MFCD17170303
  • Cas Number : 1026963-05-2
  • Molecular Weight : 238.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-7-methoxy-3-nitroquinoline
Inchi Code 1S/C10H7ClN2O3/c1-16-6-2-3-7-8(4-6)12-5-9(10(7)11)13(14)15/h2-5H,1H3
Inchi Key HOCSOOBRPRKXSA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128537
Availability details for
Quantity
  • Availability
  • Add to Cart