ENA390552926 Enamine

methyl 5-(2-bromophenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD12400790
  • Cas Number : 1035235-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126682
Availability details for
Quantity
  • Availability
  • Add to Cart