ENA390552928 Enamine

5-bromo-N-(pyridin-4-ylmethyl)pyrimidin-2-amine

  • MDL number MFCD03646012
  • Cas Number : 886365-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126684

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126684
Availability details for
Quantity
  • Availability
  • Add to Cart