ENA390552934 Enamine

3-(3-methoxyphenyl)pyrrolidin-2-one

  • MDL number MFCD19348155
  • Cas Number : 1267032-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128547
Availability details for
Quantity
  • Availability
  • Add to Cart