ENA390552941 Enamine

1-(4-chlorophenyl)-1-(1-methyl-1H-pyrazol-3-yl)methanamine

  • MDL number MFCD21179937
  • Cas Number : 1432680-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126692
Availability details for
Quantity
  • Availability
  • Add to Cart