ENA390552947 Enamine

2-chloro-3-(difluoromethoxy)pyridine

  • MDL number MFCD13185847
  • Cas Number : 1206977-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-126701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126701
Availability details for
Quantity
  • Availability
  • Add to Cart