ENA390552948 Enamine

4-(2,2-dimethylpropyl)-1,3-thiazol-2-amine

  • MDL number MFCD20720065
  • Cas Number : 4775-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126702
Availability details for
Quantity
  • Availability
  • Add to Cart