ENA390552949 Enamine

1-cyclopropyl-2-(3-methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD07379757
  • Cas Number : 1239841-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126703

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126703
Availability details for
Quantity
  • Availability
  • Add to Cart