ENA390552954 Enamine

1-(4-aminopyridin-3-yl)-2,2,2-trifluoroethan-1-one

  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-amino-3-pyridinyl)-2,2,2-trifluoroethanone
Inchi Code 1S/C7H5F3N2O/c8-7(9,10)6(13)4-3-12-2-1-5(4)11/h1-3H,(H2,11,12)
Inchi Key YHAUUIYFFGPLRP-UHFFFAOYSA-N

Description

Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-126708
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart