ENA390552956 Enamine

1-bromo-4-(2-methanesulfonylethoxy)benzene

  • MDL number MFCD12798694
  • Cas Number : 1178649-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126710

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126710
Availability details for
Quantity
  • Availability
  • Add to Cart