ENA390552968 Enamine

N,N,3,3-tetramethylpiperazine-1-carboxamide

  • MDL number MFCD19285416
  • Cas Number : 1267462-10-1
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126725
Availability details for
Quantity
  • Availability
  • Add to Cart