ENA390552969 Enamine

4-(4-fluorophenyl)cyclohex-3-en-1-one

  • MDL number MFCD12827842
  • Cas Number : 36716-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126728
Availability details for
Quantity
  • Availability
  • Add to Cart