ENA390552970 Enamine

1-methyl-1H-1,2,3,4-tetrazol-5-amine

  • MDL number MFCD00186454
  • Cas Number : 5422-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226-227
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126760

Safety Information

signalword Warning
pcodes P210,P240,P241,P264,P280,P302+P352,P305+P351+P338,P332+P313,P337+P313,P362,P370+P378
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-126760
pictograms GHS02,GHS07
hstatements H228,H315,H319
Availability details for
Quantity
  • Availability
  • Add to Cart