ENA390552975 Enamine

N-methyl-4-sulfamoylbenzamide

  • MDL number MFCD11204530
  • Cas Number : 10518-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27015

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27015
Availability details for
Quantity
  • Availability
  • Add to Cart