ENA390552978 Enamine

methyl({1-[4-(trifluoromethyl)phenyl]ethyl})amine

  • MDL number MFCD09763629
  • Cas Number : 574731-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32167
Availability details for
Quantity
  • Availability
  • Add to Cart