ENA390552982 Enamine

benzyl(1-methoxy-2-methylpropan-2-yl)amine

  • MDL number MFCD24369128
  • Cas Number : 1432681-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126774

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126774
Availability details for
Quantity
  • Availability
  • Add to Cart