ENA390552991 Enamine

6-bromo-4-chloro-8-fluoroquinoline

  • MDL number MFCD09861346
  • Cas Number : 1019016-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67744

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67744
Availability details for
Quantity
  • Availability
  • Add to Cart