ENA390553003 Enamine

methyl 2-hydroxy-2-phenylpropanoate

  • MDL number MFCD02683214
  • Cas Number : 20731-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82333

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82333
Availability details for
Quantity
  • Availability
  • Add to Cart