ENA390553015 Enamine

2-benzyl-2H-1,2,3,4-tetrazol-5-amine

  • MDL number MFCD02689458
  • Cas Number : 31694-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88766

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88766
Availability details for
Quantity
  • Availability
  • Add to Cart