ENA390553027 Enamine

4-amino-1-benzylpiperidine-4-carbonitrile

  • MDL number MFCD09042612
  • Cas Number : 136624-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97232
Availability details for
Quantity
  • Availability
  • Add to Cart