ENA390553029 Enamine

1-(4-aminopyridin-3-yl)ethan-1-one

  • MDL number MFCD19649486
  • Cas Number : 53277-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98607
Availability details for
Quantity
  • Availability
  • Add to Cart