ENA394766829 Enamine

ethyl 3-(hydroxymethyl)cyclohexane-1-carboxylate

  • MDL number MFCD24842735
  • Cas Number : 190321-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104490
Availability details for
Quantity
  • Availability
  • Add to Cart