ENA394766834 Enamine

2-[3-(1,1,2,2,2-pentafluoroethyl)phenoxy]acetic acid

  • MDL number MFCD24842742
  • Cas Number : 1443980-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121861
Availability details for
Quantity
  • Availability
  • Add to Cart