ENA394766837 Enamine

3-bromo-1-tert-butyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD24842748
  • Cas Number : 1407966-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-168
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124226
Availability details for
Quantity
  • Availability
  • Add to Cart