ENA394766847 Enamine

5H,6H,7H,8H-imidazo[1,2-a]pyridine-2-sulfonyl chloride

  • MDL number MFCD24842759
  • Cas Number : 1443980-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-115
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-125502

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-125502
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart