ENA394766848 Enamine

5-carbamoyl-2-methylthiophene-3-sulfonyl chloride

  • MDL number MFCD24842760
  • Cas Number : 1443979-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125526

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125526
Availability details for
Quantity
  • Availability
  • Add to Cart