ENA394766852 Enamine

3-chloro-5-(chlorosulfonyl)-2,4-difluorobenzoic acid

  • MDL number MFCD24842766
  • Cas Number : 1443980-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126508

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126508
Availability details for
Quantity
  • Availability
  • Add to Cart