ENA394766853 Enamine

1-(4-fluorophenyl)-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine hydrochloride

  • MDL number MFCD24842767
  • Cas Number : 1443982-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126717
Availability details for
Quantity
  • Availability
  • Add to Cart