ENA394766854 Enamine

4,4-difluoro-1,2,3,4-tetrahydroisoquinoline-1,3-dione

  • MDL number MFCD24842768
  • Cas Number : 1443981-83-6
  • Molecular Weight : 197.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,4-difluoroisoquinoline-1,3(2H,4H)-dione
Inchi Code 1S/C9H5F2NO2/c10-9(11)6-4-2-1-3-5(6)7(13)12-8(9)14/h1-4H,(H,12,13,14)
Inchi Key CNHMPGXYLZDVCT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126812

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-126812
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart