ENA394766856 Enamine

4-bromo-5-fluoro-2-methoxybenzene-1-sulfonamide

  • MDL number MFCD24842770
  • Cas Number : 1443981-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126833

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126833
Availability details for
Quantity
  • Availability
  • Add to Cart