ENA394766858 Enamine

[2-(propan-2-yloxy)ethyl]hydrazine dihydrochloride

  • MDL number MFCD24842772
  • Cas Number : 1443980-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126918
Availability details for
Quantity
  • Availability
  • Add to Cart