ENA394766859 Enamine

1-(2-aminoethyl)-5-(trifluoromethyl)-1,2-dihydropyridin-2-one hydrochloride

  • MDL number MFCD24842773
  • Cas Number : 1443981-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127044

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127044
Availability details for
Quantity
  • Availability
  • Add to Cart