ENA394766861 Enamine

(1S)-1-(1,3-thiazol-5-yl)ethan-1-ol hydrochloride

  • MDL number MFCD24842776
  • Cas Number : 1443931-85-8
  • Molecular Weight : 165.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127406
Availability details for
Quantity
  • Availability
  • Add to Cart