ENA394766862 Enamine

ethyl 4-methyl-2-(methylamino)-1,3-oxazole-5-carboxylate

  • MDL number MFCD24842778
  • Cas Number : 1443980-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127484
Availability details for
Quantity
  • Availability
  • Add to Cart