ENA394766866 Enamine

2-[(2-methoxyethyl)amino]-2-methylpropanoic acid hydrochloride

  • MDL number MFCD24842783
  • Cas Number : 1443981-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127728
Availability details for
Quantity
  • Availability
  • Add to Cart