ENA394766877 Enamine

4-methyl-5-oxo-2,3,4,5-tetrahydro-1,4-benzoxazepine-7-sulfonamide

  • MDL number MFCD24842794
  • Cas Number : 1443980-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127825
Availability details for
Quantity
  • Availability
  • Add to Cart